Learn More
Sigma Aldrich Nanofabtx(tm) Microfluidic - Micro Hardware Kit For Synthesis Of 10-30 Um Particles
Catalog No. 9118791EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. 91-187-91EA
Supplier: Sigma Aldrich 9118791EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Parameters enable facile synthesis of monodispersed particles with minimum batch-to-batch variance. Microfluidics allows scalable synthesis of particles, and rapid formulation screening.This kit was developed and tested in partnership with Dolomite Microfluidics. For compatible pumps and microfluidic systems, please visit Dolomite for more information.NANOFABTX is a trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 Ea.
Synonym Microfluidic kit; Microparticle; NanoFabTx(TM) device kit; Nanoformulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.