Learn More
Sigma Aldrich Nanofabtx(tm) Microfluidic - Micro Hardware Kit For Synthesis Of 1-5 Um Particles
Click to view available options
Quantity:
1 Ea.
Description
Parameters enable facile synthesis of monodispersed particles with minimum batch-to-batch variance. Microfluidics allows scalable synthesis of particles, and rapid formulation screening.This kit was developed and tested in partnership with Dolomite Microfluidics. For compatible pumps and microfluidic systems, Please visit: Dolomite for more informationNANOFABTX is a trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| Quantity | 1 Ea. |
| Synonym | Microfluidic kit; Microparticle; NanoFabTx(TM) device kit; Nanoformulation |