Learn More
Sigma Aldrich Nanofabtx(tm) Microfluidic - Micro Hardware Kit For Synthesis Of 10-30 Um Particles
Catalog No. 502908618
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-290-8618 1 Ea.
1 options
Catalog No. 50-290-8618 Supplier Sigma Aldrich Supplier No. 9118791EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Parameters enable facile synthesis of monodispersed particles with minimum batch-to-batch variance. Microfluidics allows scalable synthesis of particles, and rapid formulation screening.This kit was developed and tested in partnership with Dolomite Microfluidics. For compatible pumps and microfluidic systems, please visit Dolomite for more information.NANOFABTX is a trademark of Sigma-Aldrich Co. LLC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Synonym Microfluidic kit; Microparticle; NanoFabTx(TM) device kit; Nanoformulation