Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nanog Antibody (1E6C4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | Nanog |
---|---|
Clone | 1E6C4 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP147427SS
![]() |
Novus Biologicals
NBP147427SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP147427
![]() |
Novus Biologicals
NBP147427 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Nanog | |
1.0 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
NANOG | |
IgG1 | |
Protein G purified | |
35 kDa |
1E6C4 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
Q9H9S0 | |
79923 | |
Purified recombinant fragment of human Nanog (amino acids 20-166) expressed in E. coli. [UniProt# Q9H9S0] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title