Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nanog Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP213177JF549
Description
Nanog Polyclonal specifically detects Nanog in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Nanog | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NANOG | |
A synthetic peptide made to a C-terminal region of the mouse Nanog protein (within residues 1-50). [Swiss-Prot Q80Z64 ] | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Cellular Markers, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Markers, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction