Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nanog Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100588PCP
Description
Nanog Polyclonal specifically detects Nanog in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Nanog | |
Polyclonal | |
Flow Cytometry | |
FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB100-588 is specific for the Nanog protein. | |
Store at 4C in the dark. |
Flow Cytometry | |
PerCP | |
PBS with 0.05% Sodium Azide | |
NANOG | |
A synthetic peptide which maps to a region between residues 250 and the C-terminus (residue 305) of mouse Nanog homeobox using the numbering given in TrEMBL entry Q80Z64 (GeneID 71950). | |
100 μL | |
Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
79923 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction