Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog Monoclonal Antibody (23D2-3C6), DyLight™ 550, Invitrogen™

Catalog No. MA1017D550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1017D550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1017D550 Supplier Thermo Scientific Supplier No. MA1017D550

Mouse Monoclonal Antibody

MA1-017-D550 has been successfully used in ICC/IF applications with human samples. MA1-017-D550 can be used for immunofluoresence analysis of Nanog in human iPSCs.

Nanog Monoclonal antibody specifically detects Nanog in Human samples. It is validated for ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nanog
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 23D2-3C6
Concentration 1 mg/mL
Conjugate DyLight 550
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene NANOG
Gene Accession No. Q9H9S0
Gene Alias 2410002E02Rik; early embryo specific expression NK family; early embryo specific expression NK-type homeobox protein; Ecat4; ENK; ES cell-associated protein 4; FLJ12581; HGNC:20857; hNanog; Homeobox protein NANOG; homeobox transcription factor Nanog; homeobox transcription factor Nanog-delta 48; Nanog; Nanog homeobox
Gene Symbols NANOG
Host Species Mouse
Immunogen Full-length human recombinant protein expressed in bacteria
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79923
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.