Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANOG Mouse anti-Human, Clone: 60CT77.1.1, Abnova™

Catalog No. 89934648 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-648 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-648 Supplier Abnova Corporation Supplier No. MAB12279
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human NANOG.

Specifications

Antigen NANOG
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 60CT77.1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human NANOG.
Dilution Immunohistochemistry (1:10-1:50) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Symbols NANOG
Host Species Mouse
Immunogen Recombinant protein corresponding to human NANOG.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79923
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.