Learn More
Drummond Scientific Nanoject III Programmable Nanoliter Injector
Catalog No. 13681460
Change view
Click to view available options
Electrical Requirements:
100/240 V 50/60 Hz
Catalog No. Electrical Requirements
13-681-460 100/240 V 50/60 Hz
1 options
Catalog No. 13-681-460 Supplier Drummond Scientific Supplier No. 3000207
Add to Cart
Add to Cart
Request a Quote
User-friendly programmable touchscreen operation, easy positive micropipet attachments, proven hydraulic technology, universal power source, ability to create up to 8 different programs
  • The  Drummond Programmable Nanoject III Nanoliter Injector offers a number of state-of-the-art features to provide easy setup and operation as well as more accurate injection volumes down to 0.6 nanoliters.
  • The Nanoject III does not require any O-rings to secure the micropipet.
  • Operational Modes enable the user to program the unit to do single manual injections, as well as multiple injection cycle recipes.
  • Program input and execution plus the user's ability to select the injection rate widens the potential range of applications for the Nanoject III.
  • The Universal Power Source provides adapter plugs to enable the unit to be used in the US, UK, Europe, and Australia simply by inserting the proper plug.
Specifications
Electrical Requirements 100/240 V 50/60 Hz
Includes Control box & power source, Injector head, Injector head cable, 100 pcs., 3.5 in. glass capillaries, 100 pcs., 7 in. glass capillaries, Backfilling needle, Universal adapter