Learn More
Promega NanoLuc-MAPK4 Fusion Vector, 20microg
Catalog No. PRNV1681 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRNV1681 20 μg
1 options
Catalog No. PRNV1681 Supplier Promega Supplier No. NV1681
Add to Cart
Add to Cart
The NanoLuc Kinase Fusion Vectors contains the coding region of the indicated human kinase fused to either the C- or the N-terminus of NanoLuc luciferase. Designed for use with the NanoBRET Target Engagement (TE) Intracellular Kinase Assay.
  • Kinase Fusion Vector for NanoBRET™ Target Engagement
  • Transfection−ready DNA to express full−length MAPK4 kinase fused to NanoLuc(R) luciferase
  • Measures kinase target engagement in live cells
  • Use with NanoBRET™ TE Intracellular Kinase Assay, K−5
Specifications
Quantity 20 μg
Storage Requirements -30°C to -10°C