Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB10026100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10026100 100 μg
MAB10026SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10026100 Supplier R&D Systems Supplier No. MAB10026100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Luciferase Monoclonal specifically detects Luciferase in Canine∣Cercopithecus aethiops (African Green Monkey)∣Human∣Mouse∣Multi-species∣Zebrafish samples. It is validated for Immunocytochemistry∣Western Blot.

Specifications

Antigen Luciferase
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 965808
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Mouse
Immunogen Synthetic peptide of NanoLuc™ (Nluc) Luciferase
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. Detects NanoLuc™ (Nluc) Luciferase and Large BiT in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Canine, African Green Monkey, Human, Mouse, Multi-species, Zebrafish
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.