Description
- Kinase Fusion Vector for NanoBRET™ Target Engagement
- Transfection−ready DNA to express full−length TBK1 kinase fused to NanoLuc(R) luciferase
- Measures kinase target engagement in live cells
- Use with NanoBRET™ TE Intracellular Kinase Assay, K−5
Specifications
Specifications
| Quantity | 20 μg |
| Storage Requirements | -30°C to -10°C |