Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NANOS1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298706JF646
Description
NANOS1 Polyclonal antibody specifically detects NANOS1 in Human samples. It is validated for Western BlotSpecifications
NANOS1 | |
Polyclonal | |
Western Blot | |
nanos homolog 1, nanos homolog 1 (Drosophila), NOS-1, NOS1EC_Rep1a | |
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human NANOS1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
340719 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction