Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NANOS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520557
Description
A suggested positive control is rat brain tissue lysate. PA5-20557 can be used with blocking peptide PEP-0677.
NANOS1 may regulate translation of specific mRNAs by forming a complex with PUM2 that associates with the 3'-UTR of mRNA targets.
Specifications
| NANOS1 | |
| Polyclonal | |
| Unconjugated | |
| NANOS1 | |
| EC_Rep1a; nanos 1; nanos C2HC-type zinc finger 1; nanos homolog 1; nanos homolog 1 (Drosophila); Nanos1; Nos; NOS1; NOS-1; RP11-498J9.3; SPGF12; ZC2HC12A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 332397, 340719, 365494 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q80WY3, Q8WY41 | |
| NANOS1 | |
| A 14 amino acid peptide from near the center of human Nanos1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction