Learn More
Invitrogen™ NANOS1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee.
Specifications
Specifications
| Antigen | NANOS1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | NANOS1 |
| Gene Accession No. | Q8WY41 |
| Gene Alias | EC_Rep1a; nanos 1; nanos C2HC-type zinc finger 1; nanos homolog 1; nanos homolog 1 (Drosophila); Nanos1; Nos; NOS1; NOS-1; RP11-498J9.3; SPGF12; ZC2HC12A |
| Gene Symbols | NANOS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.