Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nanos3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176919
Description
Nanos3 Polyclonal specifically detects Nanos3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Nanos3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
MGC120114, nanos homolog 3, nanos homolog 3 (Drosophila), NOS-3, NOS3NANOS1L | |
Rabbit | |
20 kDa | |
0.1 mg | |
Primary | |
This Nanos3 antibody will not cross-react with either Nanos 1 or Nanos2. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
P60323 | |
NANOS3 | |
Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the human Nanos3. The immunogen is located within the last 50 amino acids of Nanos3. Amino Acid Squence: CKLVRPDKAKQDTGHR | |
Affinity Purified | |
RUO | |
342977 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction