Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NANOS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520537
Description
A suggested positive control is human brain tissue lysate. PA5-20537 can be used with blocking peptide PEP-0657.
Nanos is a zinc-finger containing, RNA-binding protein that has been implicated in germ cell development in both invertebrates and vertebrates. In Drosophila, Nanos represses apoptosis during development to ensure proper germ-line development. Unlike Nanos1 whose expression in mice is dispensable, the Nanos2 and Nanos3 proteins are required for germ cell development. Nanos2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while Nanos3-null PGCs are lost during the migration stage regardless of sex. Nanos2 and Nanos3 have distinct expression patterns during embryo development, suggesting that these two proteins do not have redundant functions. However, expression of Nanos2 can at least partially replace Nanos3 function in a Nanos3-null background. Nanos3 expression can not rescue Nanos2-null defects.
Specifications
NANOS3 | |
Polyclonal | |
Unconjugated | |
NANOS3 | |
nanos C2HC-type zinc finger 3; nanos homolog 3; nanos homolog 3 (Drosophila); NANOS1L; NANOS3; NOS3; NOS-3; ZC2HC12C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
244551, 288909, 342977 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P60323, P60324 | |
NANOS3 | |
A 16 amino acid peptide near the carboxy terminus of the human Nanos3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction