Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NANOS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564081
Description
Immunogen sequence: LAHLVRALSG KEGPETRLSP QPEPEPMLEP VSALEPMPAP ESVPVPGPKD QKRSLESSPA PERLCSFCKH NG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.
Nanos is a zinc-finger containing, RNA-binding protein that has been implicated in germ cell development in both invertebrates and vertebrates. In Drosophila, Nanos represses apoptosis during development to ensure proper germ-line development. Unlike Nanos1 whose expression in mice is dispensable, the Nanos2 and Nanos3 proteins are required for germ cell development. Nanos2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while Nanos3-null PGCs are lost during the migration stage regardless of sex. Nanos2 and Nanos3 have distinct expression patterns during embryo development, suggesting that these two proteins do not have redundant functions. However, expression of Nanos2 can at least partially replace Nanos3 function in a Nanos3-null background. Nanos3 expression can not rescue Nanos2-null defects.
Specifications
NANOS3 | |
Polyclonal | |
Unconjugated | |
NANOS3 | |
nanos C2HC-type zinc finger 3; nanos homolog 3; nanos homolog 3 (Drosophila); NANOS1L; NANOS3; NOS3; NOS-3; ZC2HC12C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
342977 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P60323 | |
NANOS3 | |
Recombinant protein corresponding to Human NANOS3. Recombinant protein control fragment (Product #RP-103481). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction