Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Nanotrap Microbiome A Particles
Shop All Streck Laboratories Products

Click to view available options
Capacity:
10 mL
30 mL
Description
- Utilize affinity capture and enrichment to attract and bind low abundance viruses.
- Can be used with clinical swab samples, saliva, urine, wastewater, food wash and blackwater.
- Compatible with PCR, sequencing or immunoassay analysis methods.
- Designed to capture and concentrate a variety of microbes.
- Increase sensitivity of existing SARS-CoV-2 assay by capturing and concentrating whole virus.
- Nanotrap Enhancement Reagents improve microbe detection in wastewater samples by at least 1-2 Ct values.
Specifications
Specifications
| Capacity | 10 mL |
| Isolation Technology | Magnetic Particles |
| Sample Type | Wastewater |
| Sequencing Type | RT-PCR and Digital PCR Sequencing |
| Product Type | Microbiome A Particles |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction