Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NANP Antibody (OTI4D11), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272870MFV610
Description
NANP Monoclonal antibody specifically detects NANP in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
NANP | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI4D11 | |
Flow Cytometry | |
C20orf147, chromosome 20 open reading frame 147, dJ694B14.3, EC 3.1.3.29, haloacid dehalogenase-like hydrolase domain containing 4, Haloacid dehalogenase-like hydrolase domain-containing protein 4, HDHD4Neu5Ac-9-Pase, MGC26833, N-acetylneuraminic acid phosphatase, N-acylneuraminate-9-phosphatase | |
Full length human recombinant protein of human NANP(NP_689880) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
140838 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction