Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANP Monoclonal Antibody (OTI4D5), TrueMAB™, OriGene
SDP

Catalog No. 502540261
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0261 30 μL
50-167-5205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0261 Supplier OriGene Supplier No. TA504547S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NANP Monoclonal antibody specifically detects NANP in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NANP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DNAAF1
Gene Accession No. Q6AYH9, Q8NEP3, Q9D2H9
Gene Alias 1600031M04Rik; C20orf147; dJ694B14.3; haloacid dehalogenase-like hydrolase domain containing 4; haloacid dehalogenase-like hydrolase domain-containing protein 4; Hdhd4; hypothetical protein LOC553795; N-acetylneuraminic acid phosphatase; N-acylneuraminate-9-phosphatase; NANP; nanp {ECO:0000250; Neu5Ac-9-Pase; RGD1306009; UniProtKB:Q9CPT3}; zgc:111947
Host Species Mouse
Immunogen Full length human recombinant protein of human LRRC50 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123872, 361419, 479628, 68270
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.