Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NANP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532940
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532940 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532940 Supplier Invitrogen™ Supplier No. PA532940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Dog, Hamster, Panda, Rabbit (100%); Marmoset, Bovine, Elephant, Pig, Opossum, Turkey, Chicken, Platypus (94%); Bat (88%); Lizard (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

NANP (N-Acetylneuraminic Acid Phosphatase) is a Protein Coding gene. Diseases associated with NANP includeParasitic Protozoa Infectious DiseaseandIntracranial Abscess. Among its related pathways areMetabolism of proteinsandSynthesis of substrates in N-glycan biosynthesis. Gene Ontology (GO) annotations related to this gene includehydrolase activityandN-acylneuraminate-9-phosphatase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NANP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NANP
Gene Accession No. Q5M969, Q8TBE9, Q9CPT3
Gene Alias 1600031M04Rik; C20orf147; dJ694B14.3; haloacid dehalogenase-like hydrolase domain containing 4; haloacid dehalogenase-like hydrolase domain-containing protein 4; Hdhd4; hypothetical protein LOC553795; N-acetylneuraminic acid phosphatase; N-acylneuraminate-9-phosphatase; NANP; nanp {ECO:0000250; Neu5Ac-9-Pase; RGD1306009; UniProtKB:Q9CPT3}; zgc:111947
Gene Symbols NANP
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human NANP.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100341646, 100764540, 140838, 311530, 608182, 67311
Target Species Human, Mouse, Rat, Canine, Rabbit, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.