Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NANS Antibody (1G6), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197938AF350
Description
NANS Monoclonal specifically detects NANS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NANS | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
NANS | |
Recombinant human NANS (1-359aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1G6 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.5.1.56, EC 2.5.1.57, N-acetylneuraminate synthase, N-acetylneuraminate-9-phosphate synthase, N-acetylneuraminic acid phosphate synthase, N-acetylneuraminic acid synthasesialic acid phosphate synthase, SASsialic acid synthase | |
Mouse | |
Protein G purified | |
metabolism, Signal Transduction | |
54187.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction