Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NANS Antibody (1G6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197938B
Description
NANS Monoclonal specifically detects NANS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NANS | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
NANS | |
Recombinant human NANS (1-359aa) purified from E. coli | |
100 μL | |
metabolism, Signal Transduction | |
54187 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1G6 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.5.1.56, EC 2.5.1.57, N-acetylneuraminate synthase, N-acetylneuraminate-9-phosphate synthase, N-acetylneuraminic acid phosphate synthase, N-acetylneuraminic acid synthasesialic acid phosphate synthase, SASsialic acid synthase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human NANS mAb, clone 1G6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NANS protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction