Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NANS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531140
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (84%), Pig (98%), Chicken (93%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme that functions in the biosynthetic pathways of sialic acids. In vitro, the encoded protein uses N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN), respectively; however, it exhibits much higher activity toward the Neu5Ac phosphate product. In insect cells, expression of this gene results in Neu5Ac and KDN production. This gene is related to the E. coli sialic acid synthase gene neuB, and it can partially restore sialic acid synthase activity in an E. coli neuB-negative mutant.
Specifications
| NANS | |
| Polyclonal | |
| Unconjugated | |
| NANS | |
| 4632418E04Rik; BB056474; epididymis secretory protein Li 100; HEL-S-100; N-acetylneuraminate synthase; N-acetylneuraminate-9-phosphate synthase; N-acetylneuraminic acid 9-phosphate synthetase; N-acetylneuraminic acid phosphate synthase; N-acetylneuraminic acid synthase; N-acetylneuraminic acid synthase (sialic acid synthase); Nans; RP11-404F11.3; SAS; SEMDG; sialic acid phosphate synthase; sialic acid synthase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NR45 | |
| NANS | |
| Recombinant protein encompassing a sequence within the center region of human NANS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction