Learn More
Description
Specifications
Specifications
| Antigen | NAP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SR2283 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Serum 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | CG5330, Dmel\CG5330, dNap, dNap1, dNAP-1, MAP3K7IP3, MGC45404, mitogen-activated protein kinase kinase kinase 7 interacting protein 3, Mitogen-activated protein kinase kinase kinase 7-interacting protein 3, Nap-1, NAP1, NF-kappa-B-activating protein 1, NFkB activating protein 1, p56/dCAF-4, TAB-3, TAK1-binding protein 3, TGF-beta activated kinase 1/MAP3K7 binding protein 3, TGF-beta-activated kinase 1 and MAP3K7-binding protein 3, TGF-beta-activated kinase 1-binding protein 3 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human NAP1 (Uniprot #: Q8N5C8) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
