Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAP1 Rabbit anti-Human, Mouse, Clone: 3Y6T7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NAP1 |
---|---|
Clone | 3Y6T7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NAP1 Monoclonal antibody specifically detects NAP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
NAP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Immunology, Innate Immunity | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
257397 | |
IgG | |
Affinity purified |
3Y6T7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CG5330, Dmel\CG5330, dNap, dNap1, dNAP-1, MAP3K7IP3, MGC45404, mitogen-activated protein kinase kinase kinase 7 interacting protein 3, Mitogen-activated protein kinase kinase kinase 7-interacting protein 3, Nap-1, NAP1, NF-kappa-B-activating protein 1, NFkB activating protein 1, p56/dCAF-4, TAB-3, TAK1-binding protein 3, TGF-beta activated kinase 1/MAP3K7 binding protein 3, TGF-beta-activated kinase 1 and MAP3K7-binding protein 3, TGF-beta-activated kinase 1-binding protein 3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NAP1 (Q8N5C8). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title