Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAP1L1 Antibody (SR1910), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32215750UL
Description
NAP1L1 Monoclonal antibody specifically detects NAP1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
NAP1L1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1910 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
hNRP, HSP22-like protein interacting protein, MGC23410, MGC8688, NAP1, NAP-1 related protein, NAP1LFLJ16112, NRPNAP-1-related protein, nucleosome assembly protein 1-like 1 | |
A synthesized peptide derived from human NAP1L1 (Uniprot #: P55209) | |
50 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
4673 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction