Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAP1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588173 Supplier Invitrogen™ Supplier No. PA588173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADIDNKEQS ELDQDLDDVE EVEEEETGEE TKLKARQLTV QMMQNPQILA ALQERLDGLV ETPTGYIESL PRVVKRRVNA LKNLQVKCAQ IEAKFYEEVH DLERKYAVLY QPLFDKRFEI INAIYEPTEE ECEWKPDEED EISEELKEKA KIEDEKKDEE KEDPKGIPEF; Positive Samples: HeLa, A357, LO2; Cellular Location: Melanosome, Nucleus.

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Nap1l1
Gene Accession No. P28656, P55209
Gene Alias AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1
Gene Symbols Nap1l1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human NAP1L1 (NP_0045281).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4673, 53605
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.