Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAP1L1 Rabbit anti-Human, Mouse, Rat, Clone: 4W8B6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | NAP1L1 |
|---|---|
| Clone | 4W8B6 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
NAP1L1 Monoclonal antibody specifically detects NAP1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| NAP1L1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4673 | |
| IgG | |
| Affinity purified |
| 4W8B6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hNRP, HSP22-like protein interacting protein, MGC23410, MGC8688, NAP1, NAP-1 related protein, NAP1LFLJ16112, NRPNAP-1-related protein, nucleosome assembly protein 1-like 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NAP1L1 (P55209). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title