Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAP1L1 Recombinant Rabbit Monoclonal Antibody (4W8B6)
GREENER_CHOICE

Catalog No. PIMA542881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542881 Supplier Invitrogen™ Supplier No. MA542881
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, 293T, A-431, NIH/3T3, Mouse testis, Rat lung, Rat spleen. The target is usually found in the following locations: Melanosome, Nucleus. Immunogen sequence: MADIDNKEQS ELDQDLDDVE EVEEEETGEE TKLKARQLTV QMMQNPQILA ALQERLDGLV ETPTGYIESL PRVVKRRVNA LKNLQVKCAQ IEAKFYEEVH.

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4W8B6
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nap1l1
Gene Accession No. P28656, P55209, Q9Z2G8
Gene Alias AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1
Gene Symbols Nap1l1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NAP1L1 (P55209).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4673, 53605, 89825
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.