Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAP1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570332 Supplier Invitrogen™ Supplier No. PA570332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.

Acidic protein which may be involved in interactions with other proteins or DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NAP1L2
Gene Accession No. P51860
Gene Alias Bpx; brain specific gene BPX; brain-specific protein, X-linked; Nap1l2; nucleosome assembly protein 1 like 2; nucleosome assembly protein 1-like 2
Gene Symbols NAP1L2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of NAP1L2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17954
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.