Learn More
Invitrogen™ NAP1L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114109
Description
Immunogen sequence: VDTLTPLIKK YDEPILKLLT DIKVKLSDPG EPLSFTLEFH FKPNEYFKNE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Acidic protein which may be involved in interactions with other proteins or DNA.
Specifications
| NAP1L2 | |
| Polyclonal | |
| Unconjugated | |
| NAP1L2 | |
| Bpx; brain specific gene BPX; brain-specific protein, X-linked; Nap1l2; nucleosome assembly protein 1 like 2; nucleosome assembly protein 1-like 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9ULW6 | |
| NAP1L2 | |
| Synthetic peptide directed towards the middle region of human NAP1L2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.