Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAP1L3 Antibody (OTI3G3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272875FR
Description
NAP1L3 Monoclonal specifically detects NAP1L3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NAP1L3 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
NAP1L3 | |
Recombinant protein fragment corresponding to amino acids 1-315 of human NAP1L3 (NP_004529) produced in E.coli. | |
0.1 mL | |
4675.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G3 | |
Western Blot, Immunohistochemistry | |
BNAP, MB20, MGC26312, NPL3, nucleosome assembly protein 1-like 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction