Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nap1l4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100631 Supplier Invitrogen™ Supplier No. PA5100631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NAP1L4.

NAP1L4 acts as histone chaperone in nucleosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nap1l4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NAP1L4
Gene Accession No. Q5U2Z3, Q78ZA7, Q99733
Gene Alias 2810410H14Rik; AI316776; D7Wsu30e; hNAP2; NAP1L4; NAP1L4b; NAP2; NAP-2; NAP2L; no arches-like zinc finger protein; nucleosome assembly protein 1 like 4; nucleosome assembly protein 1-like 4; nucleosome assembly protein 1-like 4b; nucleosome assembly protein 2; Unknown (protein for MGC:134260)
Gene Symbols NAP1L4
Host Species Rabbit
Immunogen A synthesized peptide derived from human NAP1L4(Accession Q99733), corresponding to amino acid residues H209-N259.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17955, 361684, 4676
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.