Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAP1L5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566150 Supplier Invitrogen™ Supplier No. PA566150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAEEVMAEGGA QGGDCDSAAG DPDSAAGQMA EEPQTPAENA PKPKNDFIES LPNSVKCR Highest antigen sequence identity to the following orthologs - mouse 71%, rat 71%.

This gene encodes a protein that shares sequence similarity to nucleosome assembly factors, but may be localized to the cytoplasm rather than the nucleus. Expression of this gene is downregulated in hepatocellular carcinomas. This gene is located within a differentially methylated region (DMR) and is imprinted and paternally expressed. There is a related pseudogene on chromosome 4. [provided by RefSeq, Nov 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L5
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAP1L5
Gene Accession No. Q96NT1
Gene Alias 1110020M21Rik; Down-regulated in liver malignancy; DRLM; MNCb-0385; NAP1L5; nucleosome assembly protein 1 like 5; nucleosome assembly protein 1-like 5
Gene Symbols NAP1L5
Host Species Rabbit
Immunogen Recombinant Human NAP1L5. Recombinant protein control fragment (Product #RP-106833).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.