Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPA Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300366F
Description
NAPA Polyclonal specifically detects NAPA in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NAPA | |
Polyclonal | |
ELISA, Immunocytochemistry/Immunofluorescence | |
alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
Produced in rabbits immunized with purified, recombinant NAPA (NP_003818.2; Met1-Arg295). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
8775 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction