Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
Antigen | NAPA |
---|---|
Dilution | Western Blot 1:500, ELISA 1:100-1:2000 |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019225
![]() |
Novus Biologicals
NBP2127520.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP21275205
![]() |
Novus Biologicals
NBP2127520.05MG |
0.05 mg |
Each for $402.00
|
|
|||||
NBP212752
![]() |
Novus Biologicals
NBP212752 |
0.1 mg |
Each for $670.50
|
|
|||||
Description
NAPA Polyclonal specifically detects NAPA in Human, Mouse, Rat, Porcine, Primate samples. It is validated for Western Blot, ELISA.Specifications
NAPA | |
ELISA, Western Blot | |
Unconjugated | |
alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
NAPA | |
IgG |
Western Blot 1:500, ELISA 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
8775 | |
The synthetic peptide taken from human SNAP protein were taken form region corresponding to the aa acid sequences (tmllri kkt iqg dee d). | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title