Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPA Rabbit anti-Human, Mouse, Rat, Clone: 7Z6F6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NAPA |
---|---|
Clone | 7Z6F6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NAPA Monoclonal antibody specifically detects NAPA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NAPA | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8775 | |
IgG | |
Affinity purified |
7Z6F6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
Recombinant fusion protein containing a sequence corresponding to amino acids 201-295 of human NAPA (P54920). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title