Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAPB Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548672

Catalog No. PA548672


Only null left
Add to Cart

Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAPB
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q9H115
NAPB
KLH conjugated synthetic peptide between 13-40 amino acids from the N-terminal region of human NAPB.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
NAPB
beta soluble NSF attachment protein; beta-soluble NSF attachment protein; beta-soluble NSF attachment protein {ECO:0000250; brain protein 14; brain protein I47; Brp14; b-SNAP; E161; I47; NAPB; napb {ECO:0000250; N-ethylmaleimide sensitive fusion protein attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250; N-ethylmaleimide-sensitive factor attachment protein, beta; NSF attachment protein beta; Snapb; SNAP-beta; SNAP-beta {ECO:0000250; SNARE; UniProtKB:P28663}
Rabbit
Antigen affinity chromatography, Protein A
RUO
63908
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.