Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116807 Supplier Invitrogen™ Supplier No. PA5116807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse eye, Rat brain Immunogen sequence: MDNAGKEREA VQLMAEAEKR VKASHSFLRG LFGGNTRIEE ACEMYTRAAN.

Required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAPB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NAPB
Gene Accession No. P28663, P85969, Q9H115
Gene Alias beta soluble NSF attachment protein; beta-soluble NSF attachment protein; beta-soluble NSF attachment protein {ECO:0000250; brain protein 14; brain protein I47; Brp14; b-SNAP; E161; I47; NAPB; napb {ECO:0000250; N-ethylmaleimide sensitive fusion protein attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250; N-ethylmaleimide-sensitive factor attachment protein, beta; NSF attachment protein beta; Snapb; SNAP-beta; SNAP-beta {ECO:0000250; SNARE; UniProtKB:P28663}
Gene Symbols NAPB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-50 of human NAPB (NP_0713631).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17957, 499903, 63908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.