Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphtha, VM and P, Spectrum™ Chemical
SDP

Catalog No. 18604741
Click to view available options
Quantity:
4 L
20 L
4 x 4 L
200 L
4 x 20 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

N1002, 8030-30-6

Spectrum™ Chemical Naphtha, VM and P

Chemical Identifiers

CAS 8030-30-6

Specifications

CAS Max % 1
Quantity 20 L
Packaging Steel Can
UN Number UN1268
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.