Learn More
Naphthalene-1-sulfonic acid sodium salt, 98% (dry wt.), water <3.5%
Description
Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 299°C to 301°C |
Quantity | 5 g |
Beilstein | 3639038 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 230.22 |
Percent Purity | 99% |
Assay | (dry wt.), water <2% |
Chemical Name or Material | Naphthalene-1-sulfonic acid sodium salt |
Safety and Handling
missing translation for 'healthHazard1' | H319 |
missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
missing translation for 'healthHazard3' | P264b-P280i-P305+P351+P338 |
EINECSNumber | 204-976-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.