Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene, 99%

Catalog No. p-7050023
Click to view available options
Quantity:
50 g
500 g
2 kg

Formerly important in dyestuff manufacture, as a precursor to phthalic anhydride | CAS: 91-20-3 | C10H8 | 128.17 g/mol

Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 80°C to 82°C
Density 0.9628 g/mL
Boiling Point 218°C
Flash Point 78°C (172°F)
Odor Aromatic
Refractive Index 1.5821
Quantity 50 g
UN Number UN1334
Beilstein 1421310
Merck Index 14,6370
Solubility Information Soluble in benzene,toluene,chloroform,CS2,ether; Insoluble in water; Soluble in alcohols,liquid ammonia,benzene,carbon tetrachloride,carbon disulfide,toluene,aniline,acetic acid,chloroform and butyric acid; Insoluble in water.
Formula Weight 128.17
Percent Purity 99.6%
Physical Form Crystalline
Chemical Name or Material Naphthalene
Show More Show Less
Hazard Category H228-H302-H350
Hazard Statement GHS H Statement
H228-H351-H302
Flammable solid.
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: NAPHTHALENE, REFINED
EINECSNumber 202-049-5
RTECSNumber QJ0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.