Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene-d8, for NMR, +98% atom D, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1146-65-2
C10H8
136.223
MFCD00001743
UFWIBTONFRDIAS-PGRXLJNUSA-N
naphthalene-d8,2h8 naphthalene,perdeuteronaphthalene,naphthalene d8,perdeuterated naphthalene,2 h? naphthalene,naphthalene-d8, analytical standard,naphthalene-d8 >98.0 atom % d,naphthalene d8 10 microg/ml in cyclohexane
92148
1,2,3,4,5,6,7,8-octadeuterionaphthalene
C1=CC=C2C=CC=CC2=C1
1146-65-2
C10H8
136.223
MFCD00001743
UFWIBTONFRDIAS-PGRXLJNUSA-N
naphthalene-d8,2h8 naphthalene,perdeuteronaphthalene,naphthalene d8,perdeuterated naphthalene,2 h? naphthalene,naphthalene-d8, analytical standard,naphthalene-d8 >98.0 atom % d,naphthalene d8 10 microg/ml in cyclohexane
92148
1,2,3,4,5,6,7,8-octadeuterionaphthalene
C1=CC=C2C=CC=CC2=C1
Deuterium (D) 98.0% min.
Quantity 1g
Packaging Glass bottle
Color White
Melting Point 81.0°C to 83.0°C
Linear Formula C10D8
Flash Point 78°C
Formula Weight 136.22
Physical Form Crystals or Crystalline Powder
Grade NMR
Chemical Name or Material Naphthalene-d8, For NMR
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning