Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Naphthoflavone 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6051-87-2
C19H12O2
272.29
MFCD00004986
OUGIDAPQYNCXRA-UHFFFAOYSA-N
beta-naphthoflavone,5,6-benzoflavone,beta-nf,3-phenyl-1h-naphtho 2,1-b pyran-1-one,3-phenyl-1h-benzo f chromen-1-one,1h-naphtho 2,1-b pyran-1-one, 3-phenyl,unii-1bt0256y8o,3-phenylbenzo f chromen-1-one,ccris 3262,.beta.-naphthoflavone
2361
CHEBI:77013
3-phenylbenzo[f]chromen-1-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43
6051-87-2
C19H12O2
272.29
MFCD00004986
OUGIDAPQYNCXRA-UHFFFAOYSA-N
beta-naphthoflavone,5,6-benzoflavone,beta-nf,3-phenyl-1h-naphtho 2,1-b pyran-1-one,3-phenyl-1h-benzo f chromen-1-one,1h-naphtho 2,1-b pyran-1-one, 3-phenyl,unii-1bt0256y8o,3-phenylbenzo f chromen-1-one,ccris 3262,.beta.-naphthoflavone
2361
CHEBI:77013
3-phenylbenzo[f]chromen-1-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43
Infrared Spectrum Authentic
Color Beige
Melting Point 162.0°C to 164.0°C
Packaging Glass bottle
Quantity 5g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 17, I, 216
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid
Formula Weight 272.29
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material β-Naphthoflavone, 99+%
Show More Show Less

EINECSNumber : 227-958-4

RTECSNumber : QL6200000

TSCA : TSCA