Learn More
Invitrogen™ NAPRT1 Monoclonal Antibody (CL0366)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524602
Description
Immunogen sequence: LLDTYSVWRS GLPNFLAVAL ALGELGYRAV GVRLDSGDLL QQAQEIRKVF RAAAAQFQVP WLESVLIVVS NNIDEEALAR LAQEGSEVNV IGIGTSV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%. Binds to an epitope located within the peptide sequence IRKVFRAAAA as determined by overlapping synthetic peptides.
Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).
Specifications
NAPRT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6XQN6 | |
NAPRT | |
Recombinant protein corresponding to Human NAPRT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
CL0366 | |
Unconjugated | |
NAPRT | |
9130210N20Rik; FHA-HIT-interacting protein; FHIP; hypothetical protein MGC77870; Naprt; Naprt1; NAPRTase; Nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase domain containing 1; nicotinate phosphoribosyltransferase domain-containing protein 1; nicotinate phosphoribosyltransferase; LOW QUALITY PROTEIN: nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase-like protein; nicotinic acid phosphoribosyltransferase; PP3856; zgc:77870 | |
Mouse | |
Protein A | |
RUO | |
93100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.