Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPRT1 Mouse anti-Human, Mouse, Clone: 5D8H10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NAPRT1 |
---|---|
Clone | 5D8H10 |
Concentration | 1.47 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6593
![]() ![]() |
Proteintech Group Inc
661591IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).Specifications
NAPRT1 | |
1.47 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FHA HIT interacting protein, FHIP, NAPRT1, NAPRTase, PP3856 | |
NAPRT | |
IgG2a | |
Protein A | |
Antibody |
5D8H10 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q6XQN6, Q8CC86 | |
223646, 93100 | |
NAPRT1 Fusion Protein Ag4265 | |
Primary | |
-20°C | |
NAPRT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title