Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAPRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100073 Supplier Invitrogen™ Supplier No. PA5100073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NAPRT1.

Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAPRT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NAPRT
Gene Accession No. Q6XQN1, Q6XQN6, Q8CC86
Gene Alias 9130210N20Rik; FHA-HIT-interacting protein; FHIP; hypothetical protein MGC77870; Naprt; Naprt1; NAPRTase; Nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase domain containing 1; nicotinate phosphoribosyltransferase domain-containing protein 1; nicotinate phosphoribosyltransferase; LOW QUALITY PROTEIN: nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase-like protein; nicotinic acid phosphoribosyltransferase; PP3856; zgc:77870
Gene Symbols NAPRT
Host Species Rabbit
Immunogen A synthesized peptide derived from human NAPRT(Accession Q6XQN6), corresponding to amino acid residues A199-W249.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223646, 315085, 93100
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.