Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPSA Mouse anti-Human, Clone: NAPSA/1238 + NAPSA/1239, Abnova™

Catalog No. 89035082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-082 Supplier Abnova Corporation Supplier No. MAB15062
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human NAPSA.

  • High specificity
  • High reactivity

Specifications

Antigen NAPSA
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone NAPSA/1238 + NAPSA/1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human NAPSA.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (1-2 ug/mL) The optimal working dilution should be determined by th
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. O96009
Gene Alias KAP/Kdap/NAP1/NAPA/SNAPA
Gene Symbols NAPSA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 189-299 of human NAPSA.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9476
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.